Learn More
Hospidex™ SharpSafe™ Biohazard Carton Bags
Click to view available options
Capacity (Metric):
20 L
50 L
Color:
Brown
White
Unit Size:
Pack of 10
Description
- Cardboard
- Various sizes and colors
- Disposable
Specifications
Specifications
| Capacity (Metric) | 50 L |
| Color | Brown |
| Disposable | Single-use |
| Material | Cardboard |
| Length (Metric) | 50 mm |
| Product Type | Biohazard Carton Bag |