Learn More
SharpSafe™ Polypropylene Containers
Product Code. 11989017
Click to view available options
Capacity (Metric):
0.3 L
0.45 L
0.6 L
3 L
24 L
Height (Metric):
130 mm
169 mm
205 mm
221 mm
335 mm
Unit Size:
Each
Pack of 10
Pack of 100
Pack of 50
Product Code. 11989017
Brand: SharpSafe™ SHA4170
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Use yellow SharpSafe™ Polypropylene Containers for the collection of sharp waste treatment activities and instruments that are pungent, sharp and/or pointed.
  • Translucent flip-top lid and handle
  • Yellow
  • Polypropylene
  • Autoclavable
Specifications
Capacity (Metric) 0.45 L
Certifications/Compliance Complies with UN 3291 and BS 7320 standard, NF-X-30–500, NF302, A.D.R. and UN3291, TRBA 250, Quality management system: ISO 9001:2008 and ISO 13485:2003, Environmental Standard: ISO 14001:2004
Closure Type Flip Top
Closure Color Translucent
Color Yellow
Dimensions (L x W x H) 103 x 50 x 169 mm
Height (Metric) 169 mm
Length (Metric) 103 mm
Opening Size (Metric) 40 x 30 mm
Quantity 100 Pk.
Width (Metric) 50 mm
Autoclavable Autoclavable
Includes Flip top lid, handle
Material Polypropylene