Learn More
SICCO Shelf
Product Code. 10690953
Click to view available options
Weight (Metric):
1,305 g
490 g
Unit Size:
Each
Product Code. 10690953
Brand: SICCO V186055
Only null left
Add to basket
Add to basket
Star-Desiccators Vitrum (except model Horizontal) and Tower Star Desiccators
Suitable for Vitrum-Star, Auto-Vitrum-Star, Antistatic-Star and Tower-Star Desiccators (except for Vitrum Horizontal Star) including two rails.
Specifications
Product Type Desiccator Shelf
Weight (Metric) 490 g
Width (Metric) 320 mm
Depth (Metric) 258 mm