Learn More
Witeg™ Short Adapter
Join objects with different diameter connectors with this adapter. Witeg™ Short Adapter is designed with ST male and female connection joints.
44.55€ - 46.95€
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
11827750
|
Witeg™
0 540 005 |
Socket ST: 29/32 Cone ST: 19/26. |
46.45€
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
11817750
|
Witeg™
0 540 003 |
Socket ST: 29/32 Cone ST: 14/23. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
11807750
|
Witeg™
0 540 001 |
Socket ST: 19/26 Cone ST: 14/23. |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
- Short length.
- Socket and cone of varying ST.
Specifications
DIN 12257 | |
Glass |