Learn More
Dräger™ Short-Term Detector Tubes - Perchloroethylene
Product Code. 10570773
Click to view available options
Components Measured:
Perchloroethylene 0.1/a
Perchloroethylene 10/b
Perchloroethylene 2/a
Measurement Range:
0.5 to 4ppm
10 to 500ppm
2 to 300ppm
Humidity Range:
30mg/L
5 to 12mg/L
Less than25mg/L
No. of Strokes:
1/5
3
3/9
Unit Size:
Each
Pack of 10
Product Code. 10570773
Brand: Dräger™ 8101551
  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.
Specifications
Components Measured Perchloroethylene 0.1/a
Humidity Range 30mg/L
Measurement Range 0.5 to 4ppm
No. of Strokes 3/9
Quantity 10
Response Time 3 to 9 min.
Temperature Range (English) 59°F to 86°F
Temperature Range (Metric) 15°C to 30°C
compliance-icons
Product Identifier
  • Perchloroethylene 0.1/a
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.