missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Shz-1

Product Code. 17130222
Encompass_Preferred
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 17130222

Brand: STEMCELL Technologies 73422

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Inducer of cardiac differentiation

  • Shz-1 is a cell-permeable, sulphonyl hydrazone (Shz) compound that activates cardiac differentiation through induction of genes such as Nkx2.5 (Sadek et al.)
  • Promotes cardiac differentiation in mouse induced pluripotent stem cells (Quattrocelli et al.)
  • Induces cardiac differentiation of human mobilized peripheral blood mononuclear cells; these cells enhance myocardial regenerative repair when injected in a cryo-injured rat heart model (Sadek et al.)

Specifications

CAS 326886-05-9
Synonym Sulfonylhydrazone-1
Quantity 10 mg
Molecular Formula C13H11BrN2O3S
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.