Learn More
Endecotts Sieve Shaker, Octagon 200 CL, 100-240 V, 50/60 Hz
Product Code. 15727649
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
15727649 100/115/220/230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15727649 Supplier Endecotts Supplier No. OCT200CL/100240
  • ONLINE EXCLUSIVE PRICE
    5255.00€
    4768.14€ / Pack of 1
    Save 486.86€
    9% Off
View all promotions
Estimated Shipment: 21-01-2027
Add to basket
Add to basket
Sieve Shaker Oct 200 CL
  • Octagon 200 CL sieve shaker with a measuring range from 20 μm to 125 mm, featuring electromagnetic 3D sieving motion, adjustable amplitude from 0 to 3 mm with digital setting in 0.1 mm increments, closed-loop amplitude control for consistent performance, and compatibility with 100 mm and 200 mm sieves as well as 3 in. and 8 in. sieve diameters.
Specifications
Load Bearing Capacity (Metric) 15 kg
Voltage 100/115/220/230 V