Learn More
Epredia™ Signature Series Cover Glass
Shop All Epredia Products
Click to view available options
Dimensions (L x W):
40 x 24 mm
50 x 24 mm
60 x 24 mm
Unit Size:
Pack of 10
Description
Thickness 1:
- 0.13 - 0.17mm
- Case of 10 Pk. (68-72/pack)
- Available in several sizes
Specifications
Specifications
| Length (Metric) | 24 mm |
| Width (Metric) | 60 mm |
| Thickness (Metric) | #1 (0.13 to 0.16 mm) |
| Dimensions (L x W) | 60 x 24 mm |
| Quantity | Case of 10 Pk. (68-72/pack) |