Learn More
Trajan Scientific and Medical™ SilFlow™ Ferrules
Click to view available options
Type:
SilFlow Blanking Ferrules and Pins
SilFlow Ferrules 0.35 mm ID
SilFlow Ferrules 0.8 mm ID
Quantity:
10/Pk.
5/Pk.
Unit Size:
Pack of 10
Pack of 5
Description
- Enhanced GC connectivity
- Replacement ferrules and tubing available
Specifications
Specifications
| Type | SilFlow Ferrules 0.8 mm ID |
| Description | Ferrules and pins |
| For Use With (Application) | Multi-Dimensional GC |
| Quantity | 10/Pk. |