Learn More
Trajan Scientific and Medical™ SilFlow™ Replacement Fused Silica Tubing
Microfluidic platform for multi-dimensional GC.
Brand: Trajan Scientific and Medical™ 123750
Additional Details : Weight : 0.00900kg
Description
- Enhanced GC connectivity
- Replacement ferrules and tubing available
Specifications
SilFlow Replacement Fused Silica Tubing 0.363mm OD x 0.05mm ID x 2m Length | |
0.05 mm | |
2 m |
Replacement Fused Silica Tubing | |
0.363 mm |