missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica 32 to 63 μm, 60A, EcoChrom™, Not Active, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
2.5 kg
15 kg
25 kg
Unit Size:
15kg
2.5kg
25kg
Specifications
Specifications
| Quantity | 25 kg |
| Content And Storage | Store at Room Temperature. |
| Type | Silica |
| Physical Form | Powder |
| MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
| Density | 2g/cm3 at 20°C |
| CAS | 7631-86-9 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction