Learn More
Macherey-Nagel™ Silica 60
Brand:  Macherey-Nagel™ 815650.5
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12730453
Quantity Price
1 295.00€ / 5kg
6 272.49€ / 5kg
Estimated Shipment: 04-08-2021
Specifications
60 Å
5 kg
Silica 60
Silica