Learn More
Silica Gel 60 Angstrom Pore Size, 6-35 Micron, for Column Chromatography, Fisher Chemical™
Brand: Fisher Chemical S/0691/53
Additional Details : CAS : 7631-86-9 Weight : 0.50000kg
Specifications
Plastic Powder Jar | |
500 g |
Brand: Fisher Chemical S/0691/53
Additional Details : CAS : 7631-86-9 Weight : 0.50000kg
Plastic Powder Jar | |
500 g |