missing translation for 'onlineSavingsMsg'
Learn More

Silica GF-TLC, 60A, EcoChrom™, MP Biomedicals™

Catalog Number p-4589720
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
7631-86-9
O2Si
60.08
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
24261
CHEBI:30563
silanedione
O=[Si]=O
This item is not returnable. View return policy
7631-86-9
O2Si
60.08
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
24261
CHEBI:30563
silanedione
O=[Si]=O

For Chromatography; Chemical Synthesis

Color White to Gray
Physical Form Powder
Quantity 500 g
Formula Weight 60.08
Density 2g/cm3 at 20°C
Chemical Name or Material Silica, GF-TLC
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'