Learn More
BINDER™ Left Access Port (10 to 30 mm)
For Use With: BINDER units ED 23, ED 400
Brand:  BINDER™ 8012-0537
Additional Details : Weight : 0.50000kg
Product Code. 15681776
  • 505.00€ / Each
Estimated Shipment: 19-07-2024
to see stock.
Add to basket
Description
Description
  • For use on left
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Specifications
Access port with silicone plug - 30 mm, left
Access Port
Silicone Plug
BINDER Chambers ED 23, ED 400
Left
30 mm