missing translation for 'onlineSavingsMsg'
Learn More

Silicon carbide, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7058828
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
409-21-2
CSi
40.10
MFCD00049531
HBMJWWWQQXIZIP-UHFFFAOYSA-N
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
9863
CHEBI:29390
methanidylidynesilanylium
[C-]#[Si+]
This item is not returnable. View return policy
409-21-2
CSi
40.10
MFCD00049531
HBMJWWWQQXIZIP-UHFFFAOYSA-N
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
9863
CHEBI:29390
methanidylidynesilanylium
[C-]#[Si+]

Silicon carbide powder is used as an abrasive for such as grinding wheels, whetstone, grinding wheel and sand tiles. Silicon carbide is used to produce epitaxial grapheme by graphitization at high temperatures. It is also acts asthe metallurgical deoxidizer material. Four main application areas of SiC: functional ceramics, advanced refractory, abrasives and metallurgical materials. High purity single crystal, is used to manufacture semiconductors, manufacture of silicon carbide fibers. Silicon carbide is very hard, with excellent thermal conductivity, as a semiconductor and high temperature resistant to oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 2700°C
Color Black
Physical Form Powder
Quantity 500 g
Linear Formula SiC
Merck Index 14,8492
Solubility Information It is soluble in fused potassium hydroxide,and molten iron.It is insoluble in water (0.01mg/ml) & ethanol and acid.
Formula Weight 40.1
Percent Purity 99%
Odor Odorless
Density 3.16 g/mL
Assay (metals basis)
Chemical Name or Material Silicon carbide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'