Learn More
Thermo Scientific Sterilin silicon rubber sheeting lxwxh 381 x 381 x 3.2 mm
Brand:  Thermo Scientific Sterilin SR7SHT1515180
silicon rubber sheeting lxwxh 381 x 381 x 3.2 mm
Product Code. 12812913
  • ONLINE EXCLUSIVE PRICE
    248.00€
    183.00€ / Each
    Save 65.00€
    26% Off
Only null left
Estimated Shipment: 12-03-2026