Learn More
Fisherbrand™ Silicone Gasket
Product Code. 10132131
Click to view available options
For Use With (Equipment):
Fisherbrand Mini-Plus Horizontal Gel Electrophoresis Unit
Unit Size:
Each
Product Code. 10132131
Brand: Fisherbrand™ HUSG
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Accessory for Mini-Plus Horizontal Gel Electrophoresis Unit
  • Silicone gaskets
  • Lines casting tray
  • Forms leak proof seal against inner walls of running chamber when the casting tray is turned at 90°
Specifications
For Use With (Equipment) Fisherbrand Mini-Plus Horizontal Gel Electrophoresis Unit
Type Gasket
Quantity 1 Each
Length (Metric) 1 m