Learn More
Serva Electrophoresis™ Silicone Solution
Product Code. p-8011000
Encompass_Preferred
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
Each
Product Code. 11757786
Brand: Serva Electrophoresis™ 3513003
Specifications
Item Description Silicone Solution
Quantity 250 mL
Product Type Silicone Solution
compliance-icons compliance-icons
Product Identifier
  • Silicone solution SERVA for siliconizing glass and metal
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.