Learn More
Integra Biosciences™ Silicone Tubing For DOSE IT Peristaltic Pump
Click to view available options
Diameter (Metric) Inner:
3 mm
Unit Size:
25m
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 3 mm |
| Overall Length (Metric) | 3 m |
| For Use With | For DOSE IT Peristaltic Pump |
| For Use With (Application) | For wide array of liquid waste disposal tasks carried out at the laboratory bench |