missing translation for 'onlineSavingsMsg'
Learn More

Silver molybdenum oxide, 99%

Product Code. 11304607
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
This item is not returnable. View return policy

Product Code. 11304607

Brand: Thermo Scientific Alfa Aesar 039650.01

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13765-74-7

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13765-74-7
MDL Number MFCD00053384
Synonym Silver(I) molybdate

Specifications

Melting Point 483°C
Color Beige
Physical Form Powder
Quantity 0.25 g
Formula Weight 375.68
Percent Purity 99%
Odor Odorless
Sensitivity Light sensitive
Density 6.18 g/mL
Chemical Name or Material Silver molybdenum oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.