missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate concentrate, For 1L standard solution, 0.1 M AgNO3 (0.1N), Honeywell Fluka™

Catalog Number p-7122125
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
7761-88-8
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
24470
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
7761-88-8
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
24470
CHEBI:32130
[Ag+].[O-][N+]([O-])=O

for 1L standard solution, 0.1 M AgNO3 (0.1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 1 Pc.
UN Number UN3264
IUPAC Name silver(1+) nitrate
Formula Weight 169.87g/mol
Packaging Ampoule
Chemical Name or Material Silver nitrate concentrate
compliance-icons compliance-icons
Product Identifier
  • Silver nitrate concentrate
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Silver nitrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'