missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate solution, Volumetric, 0.0282 M AgNO3 (1 ml = 1 mg Cl), Honeywell Fluka™

Catalog Number p-8302129
Click to view available options
Quantity:
1 L
6 x 1 L
7761-88-8
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
24470
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
7761-88-8
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
24470
CHEBI:32130
[Ag+].[O-][N+]([O-])=O

Volumetric, 0.0282 M AgNO3 (1 ml = 1 mg Cl)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY
Quantity 1 L
UN Number UN3082
IUPAC Name silver(1+) nitrate
Formula Weight 169.87g/mol
Concentration or Composition (by Analyte or Components) 0.0282M
Chemical Name or Material Silver nitrate solution
compliance-icons
Product Identifier
  • Silver nitrate solution
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Silver nitrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'