missing translation for 'onlineSavingsMsg'
Learn More

Silver p-toluenesulfonate, 98+%

Product Code. 11399983
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11399983

Brand: Thermo Scientific Alfa Aesar L00307.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16836-95-6 | C7H8AgO3S | 280.07 g/mol

Silver p-toluenesulfonate is used to converts alkyl halides to tosylates, and benzyl selenyl chlorides into their corresponding selenyl tosylates. It used in promotion of the leaving ability of halogens. For use in the conversion of alkyl halides to alkyl tosylates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silver p-toluenesulfonate is used to converts alkyl halides to tosylates, and benzyl selenyl chlorides into their corresponding selenyl tosylates. It used in promotion of the leaving ability of halogens. For use in the conversion of alkyl halides to alkyl tosylates.

Solubility
Soluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16836-95-6
Molecular Formula C7H8AgO3S
Molecular Weight (g/mol) 280.07
MDL Number MFCD00007511
InChI Key BRYKBDMLJJLFAB-UHFFFAOYSA-N
Synonym silver p-toluenesulfonate, silver tosylate, silverp-toluenesulfonate, silver p-toluenesulphonate, silver 1+ tosylate, silver p-tosylate, benzenesulfonic acid, 4-methyl-, silver 1+ salt, p-toluenesulfonic acid silver salt, silver i 4-methylbenzenesulfonate, silver p-methylbenzenesulfonate
PubChem CID 85606
IUPAC Name silver;4-methylbenzenesulfonate
SMILES [Ag].CC1=CC=C(C=C1)S(O)(=O)=O

Specifications

Quantity 5 g
UN Number UN3077
Solubility Information Soluble in water.
Formula Weight 279.07
Percent Purity ≥98%
Odor Odorless
Sensitivity Light sensitive
Chemical Name or Material Silver p-toluenesulfonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.