missing translation for 'onlineSavingsMsg'
Learn More

Silver powder, spherical, -325 mesh, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048241
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]
This item is not returnable. View return policy
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Spherical Powder
Quantity 10 g
UN Number UN3077
Percent Purity 99.9%
Odor Characteristic
Assay (metals basis)
Chemical Name or Material Silver powder
compliance-icons
Product Identifier
  • Silver powder
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'