Learn More
Macherey Nagel Silylation reagent MSHFBAx10 ML , 1/PK701.260.110
Brand: Macherey Nagel 701260.110
Silylation reagent MSHFBAx10 ML , 1/PK701.260.110
Safety and Handling
- MSHFBA, 1x10 mL
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Supplemental information
- MIXTURE LIST-Contains : N-methyl-N-trimethylsilyl-heptafluorobutyramide