Learn More
Macherey Nagel Silylation reagent MSHFBAx10 ML , 1/PK701.260.110
Brand:  Macherey Nagel 701260.110
Silylation reagent MSHFBAx10 ML , 1/PK701.260.110
Product Code. 12424040
  • ONLINE EXCLUSIVE PRICE
    161.00€
    145.00€ / Each
    Save 16.00€
    9% Off
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • MSHFBA, 1x10 mL
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Supplemental information
  • MIXTURE LIST-Contains : N-methyl-N-trimethylsilyl-heptafluorobutyramide