Learn More
Witeg™ Single Channel Fixed Volume Microliter Pipetter
Product Code. p-7114606
Click to view available options
Volume (Metric):
100 μL
500 μL
Unit Size:
Each
Product Code. 11827760
Brand: Witeg™ 5 401 100
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Suitable for all brands of disposable tips and ergonomically designed. Witeg™ Single Channel Fixed Volume Microliter Pipetter is conformity certified and non-fatiguing.
  • Single channel.
  • Fixed volume.
  • Precision: less than or equal to +/- 0.2%.
  • Repeatability: less than or equal to +/-0.5%.
  • Matching pipette tips: Biohit™, Gilson™, Brand™.
Specifications
For Use With Disposable Tip
Number of Channels 1
Type Fixed Volume Micropipette
Volume (Metric) 100 μL
Precision 0.2uL
Repeatability 0.5uL