Learn More
Witeg™ Single Channel Fixed Volume Microliter Pipetter
Click to view available options
Volume (Metric):
100 μL
500 μL
Unit Size:
Each
Description
- Single channel.
- Fixed volume.
- Precision: less than or equal to +/- 0.2%.
- Repeatability: less than or equal to +/-0.5%.
- Matching pipette tips: Biohit™, Gilson™, Brand™.
Specifications
Specifications
| For Use With | Disposable Tip |
| Number of Channels | 1 |
| Type | Fixed Volume Micropipette |
| Volume (Metric) | 100 μL |
| Precision | 0.2uL |
| Repeatability | 0.5uL |