Learn More
BINDER™ Single Measurement Temperature and Humidity Calibration Certificate
Product Code. p-200104337 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KBF PRO Series
KBF Series
Unit Size:
Each
Product Code. 30238204
Brand: BINDER™ 80122437
Temperature and humidity measurement taken in center of chamber with certificate.
Calibration measurement taken from the center of the chamber at 25°C/60% RH, or at specified test values.
Specifications
For Use With (Equipment) KBF Series
Duration 1 Day
Product Type Calibration Certificate