Learn More
Tenak™ Single Tray Rack for Upright Freezers, Standard
Shop All TENAK Products
Click to view available options
Dimensions (L x W x H):
420 x 139 x 170 mm
420 x 139 x 280 mm
560 x 139 x 170 mm
560 x 139 x 385 mm
Unit Size:
Each
Description
- Single trays are a drawer including a lid with a handle on the front and it will fit into any freezer
- Racks can be used with all freezer brands
- Constructed of stainless steel
Specifications
Specifications
| Material | Stainless Steel |
| For Use With (Equipment) | Upright Freezers |
| Dimensions (L x W x H) | 420 x 139 x 280 mm |
| Width (Metric) | 139 mm |