Learn More
Masterflex™ Single-Use Gamma Irradiated Sampling Bottle Assembly
Masterflex™ Single-Use Gamma Irradiated Sampling Bottle Assembly safely transfers, samples, and stores biopharmaceutical products and reagents
Brand:  Masterflex™ 95644-22
Additional Details : Weight : 0.13610kg
 View more versions of this product
Product Code. 16689561
  • 110.00€ / Each
Estimated Shipment: 19-09-2024
to see stock.
Description
Description
  • Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connections
  • Assemblies are gamma irradiated - eliminates labor and validation costs associated with assembly and cleaning
Specifications
Specifications
Gamma Irradiated
250 mL