Learn More
Poulten Graf™ Single Use Luer-Tip Two-Part Syringe
Use these syringes for veterinary medicine applications. Poulten Graf™ Single Use Luer-Tip Two-Part Syringe are PVC-free and non-toxic, sterile packed and depatched in blister packs. The single-use syringes are extremely tight, and have a transparent, smooth operating cylinder.
Brand: Poulten Graf™ 7 41001
Additional Details : Weight : 0.28000kg
Description
- Produced in accordance with DIN 13098
- Available in a variety of sizes, with and without needle
Specifications
Autoclavable | |
Polypropylene | |
Luer-tip 2-part Syringe | |
100/Pk. | |
Luer Tip | |
Sterile |
CE | |
Screen printed graduation | |
Yes | |
2 to 3 mL | |
Yes |