missing translation for 'onlineSavingsMsg'
Learn More

BLMH Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human BLMH (NP_000377.1) (Ser 2-Glu 455) fused to a polyhistidinetag at the N-terminus

Marque:  Invitrogen 11333H07E250

Informations supplémentaires : Poids : 0.10000kg

 Afficher plus de versions de ce produit

Code produit. 15288535

  • 3274.20€ / 250µg


nous consulter 056 260 260
Ajouter au panier

Description

Description

Bleomycin hydrolase (BLMH), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications., or in other research applications.
  • Activity: Measured by its ability to hydrolyze Met-AMC; the specific activity is >500pmole/min./μg
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Spécification

Spécification

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
53.4kDa
5 x 50μg
Proteases
Bleomycin hydrolase
Assay Standard
E. coli
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
NME1, AWD, GAAD, NB, NBS, NDPK-A, NDPKA, nM23, nM23-H1
BLMH
Other Proteins
His Tag
Vidéos
Documents

For Research Use Only. Not for use in diagnostic procedures.