missing translation for 'onlineSavingsMsg'
Learn More

CCL17 Recombinant Mouse Protein, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the mouse CCL17 (AAD56601.1) (Met 1-Pro 93)

Brand:  Invitrogen 50046MNAB5

Additional Details : Poids : 0.01000kg

 View more versions of this product

Product Code. 15382586

  • 93.00€ / 5µg


Expédition estimée
03-11-2020
Add to basket

Description

Description

Chemokine (C-C Motif) Ligand 17 (CCL17), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Insect
Other Cytokines
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
20295
PAGE
5μg
Full Length/Recombinant
Abcd-2, Scya17, Scya17l, Tarc
CCL17
Chemokines & Receptors
Assay Standard
Lyophilized
8.2kDa
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
<1 EU/μg
Chemokine (C-C motif) ligand 17
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.