Learn More
CSF2RB, Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Trp 443) of human CSF2RB (NP_000386.1) fused to a polyhistidine-tag at the C-terminus
Brand: Invitrogen 10516H08H25
Additional Details : Weight : 0.10000kg
Description
Colony Stimulating Factor 2 Receptor, Beta, Low-Affinity (CSF2RB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human CD131 at 10μg/mL (100μL/well) can bind biotinylated human EPOR/Fc with a linear range of 0.16-4μg/mL; 2. Immobilized human CD131 at 10μg/mL (100μL/well) can bind human GM-CSF with a linear range of 0.032-4μg/mL
- Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
50kDa | |
5 x 5μg | |
Truncated/Recombinant | |
CSF2RB, CD131, CDw131, IL3RB, IL5RB | |
CSF2RB | |
Mammalian | |
≥90% by SDS-PAGE |
1439 | |
PAGE | |
Store at -5° to -30°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
<1 EU/μg | |
Colony stimulating factor 2 receptor, beta, low-affinity | |
His Tag | |
Cytokines & Receptors/CD Proteins |
For Research Use Only. Not for use in diagnostic procedures.