missing translation for 'onlineSavingsMsg'
Learn More

HA Recombinant Influenza A, Subtype H1N1 (A/Puerto Rico/8/1934) Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 528) of the influenza A hemagglutinin (A/Puerto Rico/8/1934 (H1N1)) (ABD77675.1), fused to a C-terminal polyhistidine tag

Brand:  Invitrogen 11684V08H5

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15278845

  • 86.50€ / 5µg


Estimated Shipment
21-08-2020
Add to basket

Description

Description

Hemagglutinin (HA), recombinant viral protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Assay Standard
Lyophilized
PAGE
5μg
C-Terminal
<1 EU/μg
Hemagglutinin
His Tag
Cell Structure/Organelle Proteins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
59kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Truncated/Recombinant
HA
HA
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.