missing translation for 'onlineSavingsMsg'
Learn More

LGALS1 Recombinant Human Protein, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human LGALS1 (NP_002296.1) (Met 1-Asp135)

Marque:  Invitrogen 10290HNAE5

Informations supplémentaires : Poids : 0.10000kg

 Afficher plus de versions de ce produit

Code produit. 15285085

  • 68.60€ / 5µg


Date dexpédition estimée
du fournisseur 29-10-2019
Ajouter au panier

Description

Description

Lectin, galactoside-binding, soluble, 1 (LGALS1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by its ability to agglutinate human red blood cells, the ED50 for this effect is typically 0.5-3μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Spécification

Spécification

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
15kDa
5μg
Lectins & Leptins
Lectin, galactoside-binding, soluble, 1
Assay Standard
E. coli
Lyophilized
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
LGALS1, DKFZp686E23103, GAL1, GBP
LGALS1
Other Proteins
Untagged
Vidéos
Documents

For Research Use Only. Not for use in diagnostic procedures.