missing translation for 'onlineSavingsMsg'
Learn More

MCAM (CD146) Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 559) of human MCAM precursor (NP_006491.2) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Invitrogen 10115H02H250

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13495716

  • 1465.00€ / 250µg


Estimated Shipment
15-09-2020
Add to basket

Description

Description

Melanoma cell adhesion molecule (MCAM/CD146), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of the HUVEC human umbilical vein endothelial cell line; when 3 x 104 cells/well are added to human CD146 coated plates (0.8μg/mL, 100μL/well), approximately >30 % will adhere after one hour at 37°C
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Other Proteins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
4162
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Melanoma cell adhesion molecule
IgG1 Fc Tag
CD Proteins
Assay Standard
Lyophilized
86.4kDa
5 x 50μg
Truncated/Recombinant
MCAM, CD146, MUC18
MCAM
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.