missing translation for 'onlineSavingsMsg'
Learn More

RSPO1 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™, 28.2kDa

This recombinant protein was expressed from a DNA sequence encoding the human RSPO1 (NP_001033722.1) (Met 1-Ala 263) fused to a polyhistidine tag at the C-terminus

71.15€ - 312.00€

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP11-566C13.1, CRISTIN3, FLJ40906, RSPO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15268195
View Documents
Invitrogen
11083H08H5
5μg 71.15€
5µg View alternative products
Save Up to 
»
Estimated Shipment
17-09-2020
Add to basket
 
15278195
View Documents
Invitrogen
11083H08H50
50μg 312.00€
50µg View alternative products
Save Up to 
»
Estimated Shipment
17-09-2020
Add to basket
 
Description

Description

R-Spondin 1 (RSPO1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind recombinant mouse CD36 in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
RP11-566C13.1, CRISTIN3, FLJ40906, RSPO

For Research Use Only. Not for use in diagnostic procedures.