missing translation for 'onlineSavingsMsg'
Learn More

SEMA3A Recombinant Mouse Protein, N-hIgG1-Fc Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the N-terminal fragment (Lys 26-Phe 546) of mouse SEMA3A (O08665) fused to the Fc region of human IgG1 at the N-terminus

Brand:  Invitrogen 50631M01H50

Additional Details : Poids : 0.01000kg

 View more versions of this product

Product Code. 15379066

  • 341.00€ / 50µg


Expédition estimée
03-11-2020
Add to basket

Includes: One vial of protein in suspension buffer

Description

Description

Sema Domain, Immunoglobulin Domain (Ig) Short Basic Domain, Secreted, (Semaphorin) 3A (SEMA3A), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Assay Standard
Lyophilized
87.7kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
Hsema-I, SEMA1, SemD, Semad, coll-1
SEMA3A
Other Proteins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
20346
PAGE
50μg
N-Terminal
<1 EU/μg
Sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3a
IgG1 Fc Tag
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.