missing translation for 'onlineSavingsMsg'
Learn More

TNF (TNFA) Recombinant Rat Protein, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the mature form of rat TNF-α (P 16599) (Val 80-Leu 235) fused to an initial Met at the N-terminus

Brand:  Invitrogen 80045RNAE5

Additional Details : Poids : 0.10000kg

 View more versions of this product

Product Code. 15379556

  • 89.50€ / 5µg


Expédition estimée
03-11-2020
Add to basket

Includes: One vial of lyophilized protein

Description

Description

Tumor Necrosis Factor (TNF), recombinant rat protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured in a cytotoxicity assay using L930 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D; the ED50 is typically 10-20pg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

E. coli
TNF (Tumor Necrosis Factor)
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
17.4kDa
5μg
Tnf, mgC124630, RATTNF, TNF-alpha, Tnfa
TNF
Cytokines & Receptors
Assay Standard
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Tumor necrosis factor
Untagged
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.