Learn More
RETSCH Sintered Corundum Comfort Grinding Jars
Product Code. p-8004893 Shop All Retsch Products
Click to view available options
Capacity (Metric):
50 mL
125 mL
250 mL
500 mL
Unit Size:
Each
Product Code. 10348612
Brand: RETSCH 01.462.0221
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Use these grinding jars with Retsch™ ball mills. Retsch™ Sintered Corundum Comfort Grinding Jars feature an O-type sealing for safe operation.
  • Available in a variety of sizes
  • Optional aeration covers
  • Pressure tight safety closure devices
Specifications
Product Type Comfort Grinding Jar
Capacity (Metric) 250 mL
For Use With (Equipment) For PM 100, PM 400 and S 100
Material Sintered Corundum