missing translation for 'onlineSavingsMsg'
Learn More

SIRT1/2 Inhibitor IV, Cambinol, TRC

Product Code. 30412058
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30412058

Brand: Toronto Research Chemicals TRCS4879055MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material SIRT1/2 Inhibitor IV','Cambinol
CAS 14513-15-6
Molecular Formula C21H16N2O2S
Quantity 5 mg
Synonym 2,3-Dihydro-5-[(2-hydroxy-1-naphthalenyl)methyl]-6-phenyl-2-thioxo-4(1H)-pyrimidinone, 5-[(2-Hydroxynaphthalen-1-yl)methyl]-6-phenyl-2-thioxo-2,3-dihydro-1H-pyrimidin-4-one, Cambinol, NSC 112546
InChI Formula InChI=1S/C21H16N2O2S/c24-18-11-10-13-6-4-5-9-15(13)16(18)12-17-19(14-7-2-1-3-8-14)22-21(26)23-20(17)25/h1-11,24H,12H2,(H2,22,23,25,26)
SMILES Oc1ccc2ccccc2c1CC3=C(NC(=S)NC3=O)c4ccccc4
IUPAC Name 5-[(2-hydroxynaphthalen-1-yl)methyl]-6-phenyl-2-sulfanylidene-1H-pyrimidin-4-one
Molecular Weight (g/mol) 360.43
Formula Weight 360.0932
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.