Learn More
Notrax Skystep™ Nitrile Rubber Mat with Moulded Edges
Ergonomically and economically engineered, the Skystep™ is designed to suction to the ground to prevent the mat from shifting while trapping air in its chambers. These air pockets combined with the resiliency of the rubber create an ultimate cushioning effect while keeping the mat lightweight.
Brand: Notrax 456S0034BL
Additional Details : Weight : 11.00000kg
Description
- Ergonomically and economically engineered, the Skystep™ utilizes the power of air
- Skystep™ is designed to suction to the ground to prevent the mat from shifting while trapping air in its chambers
- These air pockets combined with the resiliency of the rubber create an ultimate cushioning effect while keeping the mat lightweight
- Integrated bevelled edges on all four sides to prevent tripping on the mat
- Loose lay with no installation required
- Slip resistance rating R9 according to DIN 51130 and BG-rule BGR181
- Free of DOP, free of DMF, free of ozone-depleting substances, free of silicone and free of heavy metals
- 100% nitrile rubber compound
- Resistant to most industrial oils
Specifications
4 ft. | |
Industrial and Commercial Matting | |
90 cm |
Black | |
Nitrile Rubber | |
120 cm |