missing translation for 'onlineSavingsMsg'
Learn More

SLC6A4 Recombinant Rabbit Monoclonal Antibody (16H2L7), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen 702076

Additional Details : Weight : 0.01000kg

Product Code. 15657728

  • 470.00€ / 100µg
Estimated Shipment: 09-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is predicted to react with Monkey, Horse and Cat Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

SLC6A4 Recombinant Monoclonal antibody specifically detects SLC6A4 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.
Specifications

Specifications

SLC6A4
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P31645, Q60857, P31652
SLC6A4
Peptides corresponding to Human SLC6A4 (aa 444-462, 16-29)
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Immunocytochemistry, Western Blot
16H2L7
Unconjugated
SLC6A4
5 OCD1; 5HT transporter; 5HTT; 5-HTT; 5-HTTLPR; 5-hydroxytryptamine (serotonin) transporter; AI323329; HGNC:11050; hSERT; HTT; Na+/Cl- dependent serotonin transporter; OCD1; OTTHUMP00000163633; serotonin transporter; serotonin transporter 1; Sert; SERT1; Slc6a4; Sodium-dependent serotonin transporter; Sodium-dependent serotonin transporter (5HT transporter) (5HTT); solute carrier family 6 (neurotransmitter transporter serotonin) member 4 (5-hydroxytryptamine (serotonin) transporter); Solute carrier family 6 (neurotransmitter transporter serotonin) member 4 (5-hydroxytryptamine (serotonin) transporter); solute carrier family 6 (neurotransmitter transporter), member 4; solute carrier family 6 (neurotransmitter transporter, serotonin), member 4; solute carrier family 6 member 4; solute carrier family 6, member 4
Rabbit
Protein A
RUO
15567, 25553, 6532
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions