Learn More
Epredia™ SlideMate™ AS Slide Printer
Product Code. 15408891 Shop All Epredia Products
Click to view available options
Product Type:
Slide Printer AS Basic
Slide Printer AS Basic System
Slide Printer AS On-Demand
Slide Printer AS On-Demand Sys
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 15408891
Brand: Epredia™ A83900002
Regulatory: This product is under eco-contribution tax.
Reduce specimen identification errors while enhancing lean workflow with the Epredia™ SlideMate™ AS On-Demand Slide Printer.
The SlideMate AS is the next generation Epredia Slide Printer delivering a smaller footprint than previous models, making it even easier to fit next to the microtome. The SlideMate AS promotes single piece workflow though unique features designed specifically for on-demand printing.
  • Fully integrated color touch-screen and simple software are easy to use
  • Build-in barcode scanner reads cassette information and imports the data to the slide label
  • Connects easily to other computers or Laboratory Information Systems
  • Prints directly onto the slide, eliminating the need for hand writing or labels
TRUSTED_SUSTAINABILITY
Specifications
Product Type Slide Printer AS Basic System
Includes SlideMate AS basic printer with slide delivery system, color touchscreen, USB connection, power cord and simple software to print externally generated slide label from an image file.