Learn More
Edmund Bühler GmbH™ Sliding Plate with Combifix VKS
Affix to a multistory platform. Edmund Bühler GmbH™ Sliding Plate with Combifix VKS fastens horizontal separating funnels of different sizes to platform.
Brand: Edmund Bühler GmbH™ 0051 493
Additional Details : Weight : 0.80000kg
Description
- Fastens horizontal separating funnels of different sizes to platform
- For use with Combifix VKS or SM Universal Shakers
Specifications
Sliding Plate | |
Sliding plates with Combifix VKS A | |
9 Clamping Strips H, base plate and basic rack |
760 x 600 mm | |
Combifix VKS |