Learn More
Edmund Bühler GmbH™ Sliding Plate with Combifix SM
Affix to a multistory platform. Edmund Bühler GmbH™ Sliding Plate with Combifix SM fastens horizontal separating funnels of different sizes to platform.
630.00€ - 745.00€
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
10540102
|
Edmund Bühler GmbH™
0051 484 |
5 Clamping Strips H, additional base plate |
630.00€
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
10071051
|
Edmund Bühler GmbH™
0051 485 |
4 Clamping Strips H, 4 Clamping Strips V, additional base plate |
745.00€
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
Description
- Fastens horizontal separating funnels of different sizes to platform
- For use with Combifix VKS or SM Universal Shakers
Specifications
Sliding Plate |