missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SM68 Standard 1: Al,As,B,Ba,Be,Bi,Ca,Cd,Ce,Co,Cr,Cs,Cu,Dy,Er,Eu,Fe,Ga,Gd,Ho,In,K,La,Li,Lu,Mg,Mn,Na,Nd,Ni,P,Pb,Pr,Rb,Re,Sc,Se,Sm,Sr,Tb,Th,Tl,Tm,U,V,Y,Yb,Zn @ 100 μg/mL in 5% HNO3, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
SM68 Standard 1: Al,As,B,Ba,Be,Bi,Ca,Cd,Ce,Co,Cr,Cs,Cu,Dy,Er,Eu,Fe,Ga,Gd,Ho,In,K,La,Li,Lu,Mg,Mn,Na,Nd,Ni,P,Pb,Pr,Rb,Re,Sc,Se,Sm,Sr,Tb,Th,Tl,Tm,U,V,Y,Yb,Zn @ 100 μg/mL in 5% HNO3
Specifications
Specifications
| Chemical Name or Material | Aluminium/Arsenic/Barium/Beryllium/Bismuth/Boron/Cadmium/Caesium/Calcium/Cerium/Chromium/Cobalt/Copper/Dysprosium/Erbium/Europium/Gadolinium/Gallium/Holmium/Indium/Iron/Lanthanum/Lead/Lithium/Lutetium/Magnesium/Manganese/Neodymium/Nickel/Phosphorus/Potassium/Praseodymium/Rhenium/Rubidium/Samarium/Scandium/Selenium/Sodium/Strontium/Tantalum/Terbium/Thallium/Thorium/Uranium/Vanadium/Ytterbium/Yttrium/Zinc |
| CAS | 7429-90-5, 7440-38-2, 7440-39-3, 7440-41-7, 7440-69-9, 7440-42-8 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Aluminium/Arsenic/Barium/Beryllium/Bismuth/Boron/Cadmium/Caesium/Calcium/Cerium/Chromium/Cobalt/Copper/Dysprosium/Erbium/Europium/Gadolinium/Gallium/Holmium/Indium/Iron/Lanthanum/Lead/Lithium/Lutetium/Magnesium/Manganese/Neodymium/Nickel/Phosphorus/Potassium/Praseodymium/Rhenium/Rubidium/Samarium/Scandium/Selenium/Sodium/Strontium/Tantalum/Terbium/Thallium/Thorium/Uranium/Vanadium/Ytterbium/Yttrium/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 100 μg/mL/Arsenic 100 μg/mL/Barium 100 μg/mL/Beryllium 100 μg/mL/Bismuth 100 μg/mL/Boron 100 μg/mL/Cadmium 100 μg/mL/Caesium 100 μg/mL/Calcium 100 μg/mL/Cerium 100 μg/mL/Chromium 100 μg/mL/Cobalt 100 μg/mL/Copper 100 μg/mL/Dysprosium 100 μg/mL/Erbium 100 μg/mL/Europium 100 μg/mL/Gadolinium 100 μg/mL/Gallium 100 μg/mL/Holmium 100 μg/mL/Indium 100 μg/mL/Iron 100 μg/mL/Lanthanum 100 μg/mL/Lead 100 μg/mL/Lithium 100 μg/mL/Lutetium 100 μg/mL/Magnesium 100 μg/mL/Manganese 100 μg/mL/Neodymium 100 μg/mL/Nickel 100 μg/mL/Phosphorus 100 μg/mL/Potassium 100 μg/mL/Praseodymium 100 μg/mL/Rhenium 100 μg/mL/Rubidium 100 μg/mL/Samarium 100 μg/mL/Scandium 100 μg/mL/Selenium 100 μg/mL/Sodium 100 μg/mL/Strontium 100 μg/mL/Tantalum 100 μg/mL/Terbium 100 μg/mL/Thallium 100 μg/mL/Thorium 100 μg/mL/Uranium 100 μg/mL/Vanadium 100 μg/mL/Ytterbium 100 μg/mL/Yttrium 100 μg/mL/Zinc 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction