Learn More
LABPLAS Small Rack
Product Code. p-200102266
Click to view available options
Includes:
None
Sterile Bags
Unit Size:
Each
Pack of 3
Product Code. 17980295
Brand: LABPLAS SA53070
Only null left
Add to basket
Add to basket
Small rack designed for the safe transport or storage of sample bags.
  • Facilitates the transport and maintenance of sample bags
  • Foldable, lightweight, sturdy, easy to store and dishwasher safe
  • Ideal for sampling activities in the laboratory or in the field
  • Allows you to keep your hands free as you work with sample bags
Specifications
Includes None
Product Type Bag Rack
Autoclavable Not Autoclavable
Holds Bags