Learn More
Thermo Scientific™ Smart-Tracker™ Wireless Data Module Accessories
Maximize Thermo Scientific™ Smart-Tracker™ with additional accessories or personalize your Smart-Tracker with a series of colored bands.
Specifications
| For Use With (Application) | Wireless Monitoring Solution |
|---|---|
| For Use With (Equipment) | Smart-Tracker™ Module |
| Dimensions (L x W x H) | 0.24 x 0.63 x 1.02 in. (6 x 16 x 26 mm) [Shipping Dimensions] |
Description
Maximize Thermo Scientific™ Smart-Tracker™ with additional accessories or personalize your Smart-Tracker with a series of colored bands.
Specifications
| Wireless Monitoring Solution | |
| Smart-Tracker™ Module | |
| 0.24 x 0.63 x 1.02 in. (6 x 16 x 26 mm) [Shipping Dimensions] |